Learn More
Medchemexpress LLC Phosphate acetyltransferase | 9029-91-8 | 1 KU
SDP
Supplier:  Medchemexpress LLC HYP29431KU
Encompass_Preferred
Phosphate acetyltransferase is a transferase enzyme, frequently utilized in biochemical studies. It catalyzes the reversible transfer of the acetyl group from acetyl-P to CoA, forming acetyl-CoA and inorganic phosphate, thereby participating in acetate assimilation/dissimilation reactions.
Catalog No. 50-003-96607