Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Specifications
Chemical Name or Material | Phosphate Buffer, 0.05M |
---|---|
CAS | 7732-18-5, 64-19-7, 7778-77-0, 7664-38-2, 6153-56-6, 79-09-4 |
CAS Min % | 91.0, 5.76, 0.31, 0.26, 0.18, 0.09 |
CAS Max % | 95.0, 5.99, 0.32, 0.27, 0.19, 0.09 |
Specifications
Phosphate Buffer, 0.05M | |
7732-18-5, 64-19-7, 7778-77-0, 7664-38-2, 6153-56-6, 79-09-4 | |
91.0, 5.76, 0.31, 0.26, 0.18, 0.09 | |
95.0, 5.99, 0.32, 0.27, 0.19, 0.09 |
Safety and Handling

Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title