Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Akt1 (Ser473) Recombinant Rabbit Monoclonal Antibody (AktS473-C7), FITC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$199.80 - $507.00
Specifications
Antigen | Phospho-Akt1 (Ser473) |
---|---|
Clone | AktS473-C7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA537222
|
Thermo Scientific
MA537222 |
10 Tests |
Each for $199.80
|
|
|||||
PIMA536905
|
Thermo Scientific
MA536905 |
100 Tests |
Each for $507.00
|
|
|||||
Description
Phospho-Akt1 (Ser473) Recombinant Monoclonal antibody specifically detects Phospho-Akt1 (Ser473) in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Phospho-Akt1 (Ser473) | |
Flow Cytometry | |
FITC | |
Rabbit | |
Human, Mouse | |
P31749, P31750 | |
11651, 207 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser473 of human phospho Akt1. | |
Primary | |
4° C, store in dark | |
AKT1 |
AktS473-C7 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.20% BSA and 0.09% sodium azide | |
Akt; Akt kinase; AKT serine/threonine kinase 1; Akt/PKB; akt1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; CAKT; C-AKT; CG4006; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; dAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; DAKT1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; dPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; l(3)04226; l(3)89Bq; murine thymoma viral (v-akt) oncogene homolog 1; pAkt; P-AKT; PKB; PKB alpha; PKB beta; PKB gamma; PKB/AKT; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBG; PRKBA; protein kinase B; Protein kinase B alpha; protein kinase B-alpha; proto-oncogene c-Akt; RAC; rac protein kinase alpha; RAC protein kinase alpha RAC-PK alpha; RAC serine/threonine protein kinase; RAC serine/threonine-protein kinase; RAC-ALPHA; RAC-alpha serine/threonine-protein kinase; RacPK; RAC-PK-alpha; RAC-PK-beta; RAC-PK-gamma; related to A and C kinases; related to PKA to PKC protein kinases; related to the A and C kinases; SD10374p; serine/threonine protein kinase | |
AKT1 | |
IgG κ | |
Protein A/G | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title