Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Alpha synuclein (Tyr136) Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS5626R
Description
Alpha-Synuclein belongs to the Synuclein family, which includes beta and gamma Synuclein, and is predominantly expressed in neurons, and concentrated at synaptic terminals. Synucleins may be involved in the regulation of dopamine release and transport and the modulation of synaptic vesicle function. Alpha-Synuclein is a small, presynaptic protein that is the major nonamyloid component of the pathological inclusions characteristic of a wide range of neurodegenerative disorders, collectively known as synucleinopathies. Alpha-Synuclein is involved in the formation of SNARE complexes, and most significantly, aggregated alpha-synuclein is one of the major components found in the Lewy bodies that occur in Parkinson ft.s disease (PD) and other neurodegenerative disorders. Mutation of the alpha-synuclein gene is associated with familial forms of PD. Alpha-Synuclein is known to reduce the fibrillization of the microtubule-associated protein, tau, to bind various targets such as 14-3-3, protein kinase C, synphilin-1, Elk and Tat-binding protein 1. The alpha and beta synucleins are found primarily in brain tissue where they are seen mainly in neuronal cytosol and presynaptic terminals. The antibody specific for alpha-synuclein does not cross react with beta-synuclein and will be useful for the confirmation of Parkinson ft.s disease and Lewy body dementia and other studies for the localization and detection of alpha-synuclein. Four alternatively spliced transcripts that encode two different isoforms of the alpha-synuclein protein have been identified.Specifications
Phospho-Alpha synuclein (Tyr136) | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SNCA | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
6622 |
SNCA | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P37840 | |
Alpha synuclein; alphaSYN; Alpha-synuclein; MGC110988; MGC110988 antibody; NACP; NACP antibody; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; OTTHUMP00000218549; OTTHUMP00000218551; OTTHUMP00000218552; OTTHUMP00000218553; OTTHUMP00000218554; PARK 1; PARK 1 antibody; PARK 4; PARK 4 antibody; PARK1; PARK1 antibody; PARK4; PARK4 antibody; PD 1 antibody; PD1; PD1 antibody; SNCA; SNCA antibody; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor); SYUA | |
KLH conjugated synthetic phosphopeptide derived from human SNCA around the phosphorylation site of Tyr136 [QD (p-Y)EP]. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title