Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-APBB1 (Ser175) Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS5165R
Description
The protein encoded by this gene is a member of the Fe65 protein family. It is an adaptor protein localized in the nucleus. It interacts with the Alzheimer ft.s disease amyloid precursor protein , transcription factor CP2/LSF/LBP1 and the low-density lipoprotein receptor-related protein. APP functions as a cytosolic anchoring site that can prevent the gene product ft.s nuclear translocation. This encoded protein could play an important role in the pathogenesis of Alzheimer ft.s disease. It is thought to regulate transcription. Also it is observed to block cell cycle progression by downregulating thymidylate synthase expression. Multiple alternatively spliced transcript variants have been described for this gene but some of their full length sequence is not known.Specifications
Phospho-APBB1 (Ser175) | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
APBB1 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
322 |
APBB1 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O00213 | |
adaptor protein FE65a2; amyloid beta (A4) precursor protein-binding, family B, member 1; amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65); amyloid beta A4 precursor protein-binding family B member 1; amyloid beta precursor protein binding family B member 1; Amyloid-beta A4 precursor protein-binding family B member 1; Apbb1; FE65; MGC:9072; Protein Fe65; RIR; stat-like protein | |
KLH conjugated synthetic phosphopeptide derived from human APBB1 around the phosphorylation site of Ser175 [LS (p-S)PP]. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title