Learn More
Phospho-ASK1 (Ser83) Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS5436R
Catalog No. 50-198-6563
Add to cart
Description
Description
Mitogen-activated protein (MAP) kinase cascades are activated in response to various extracellular stimuli, including cytokines, growth factors and environmental stresses. A novel MAP kinase kinase kinase (MAPKKK) was identified and designated ASK1 (for apoptosis signal-regulating kinase 1) and MAPKKK5. ASK1 activates two different subgroups of MAPKK, MKK4 and MKK6, which in turn activates c-Jun N-terminal kinase (JNK) and p38 MAP kinase, respectively. ASK1/MAPKKK5 is activated by TNFR and Fas through the interaction with members of the TRAF family and Fas-associated protein Daxx. Overexpression of ASK1 induced apoptotic cell death and a catalytically inactive form of ASK1 inhibited TNF-a-induced apoptosis. ASK1 is expressed in variety of human and mouse tissues.
Specifications
Specifications
Phospho-ASK1 (Ser83)
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MAP3K5
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
4217
MAP3K5
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q99683
7420452D20Rik; apoptosis signal regulating kinase 1; Apoptosis signal-regulating kinase 1; ASK; ASK 1; ASK1; ASK-1; EC 2.7.11.25; kinase ASK1; M3K5; MAP/ERK kinase kinase 5; MAP3K5; MAPK/ERK kinase kinase 5; MAPKKK5; MEK kinase 5; MEKK 5; Mekk5; mitogen activated protein kinase kinase kinase 5; Mitogen-activated protein kinase kinase kinase 5; RGD1306565; RP3-325F22.4
KLH conjugated synthetic phosphopeptide derived from human ASK1 around the phosphorylation site of Ser83.
Protein A
RUO
Antibody
Polyclonal
Human