Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-B-Myb (Ser577, Ser581) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538428
Description
c-Myb is the cellular homolog of the leukemogenic avian retroviral protein v-Myc. c-Myb is expressed predomitly in immature and rapidly dividing hematopoietic cells, and cellular levels of c-Myb substantially decreases as cells reach terminal differentiation. B-Myb is expressed in a wide variety of proliferating cells, with levels accumulating during the G1 to S phase transition. A-Myb is expressed at specific times in reproductive tissues, some neural cells, and a subset of normal and neoplastic B lymphocytes.
Specifications
Phospho-B-Myb (Ser577, Ser581) | |
Polyclonal | |
Unconjugated | |
MYBL2 | |
Myb-related protein B, B Myb, Myb-like protein 2, MYBL2, BMYB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17865, 296344, 4605 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P10244, P48972 | |
MYBL2 | |
A synthetic phosphopeptide derived from human B-Myb around the phosphorylation site of Ser577/581 (R-R-SP-P-I) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction