Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Bim (Ser44, Ser104) Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA564656

Catalog No. PIPA564656


Add to cart

Description

Description

Phospho-Bim (Ser44, 104) Polyclonal Antibody detects endogenous levels of Bim only when phosphorylated at Ser44, 104.

Bim belongs to the Bcl2 family of proteins containing a Bcl2 homology domain 3 (BH3). It is proapoptotic and exerts its effects by interacting with prosurvival members of the Bcl2 family like Bcl2, BclxL and Bclw.
Specifications

Specifications

Phospho-Bim (Ser44, Ser104)
Polyclonal
Unconjugated
BCL2L11
Liquid
BCL2L11
A synthesized peptide derived from human Bim (Phospho-Ser44/104)
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O43521, O54918, O88498
Bim-A
Rabbit
IgG
100 μL
-20°C
Primary
10018, 12125, 64547
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit