Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-BRCA1 (Ser1524) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104888
Description
Antibody detects endogenous levels of BRCA1 only when phosphorylated at Serine 1524.
BRCA1 (breast and ovarian cancer susceptibility protein 1) is a nuclear phosphoprotein that plays a role in maintaining genomic stability and acts as a tumor suppressor in combination with other tumor suppressors, DNA damage sensors, and signal transducers to form a large multi-subunit protein complex known as BASC (for BRCA1 associated genome surveillance complex). BRCA1 associates with RNA polymerase II, and through the C terminal domain, also interacts with histone deacetylase complex. The BRCA1 protein thus plays a role in transcription, DNA repair of double-stranded breaks, and recombination. Mutations in the BRCA1 gene are responsible for approximately 40% of inherited breast cancers and more than 80% of inherited breast and ovarian cancers. Alternative splicing plays a role in modulating the subcellular localization and physiological function of this gene. Many alternatively spliced transcript variants, some of which are disease-associated mutations, have been described for this gene, but the full-length natures of only some of these variants has been described. A related pseudogene, which is also located on chromosome 17, has been identified.
Specifications
Phospho-BRCA1 (Ser1524) | |
Polyclonal | |
Unconjugated | |
BRCA1 | |
brca 1; Brca1; brca-1; BRCA1, DNA repair associated; BRCA1/BRCA2-containing complex, subunit 1; BRCAI; BRCC1; breast and ovarian cancer susceptibility protein 1; breast and ovarian cancer sususceptibility protein 1; breast cancer 1; breast cancer 1 early onset; breast cancer 1, early onset; breast cancer associated 1; breast cancer type 1 susceptibility protein; Breast cancer type 1 susceptibility protein homolog; BROVCA1; BROVCA12; Delta772-3095; early onset breast cancer 1; Fanconi anemia, complementation group S; FANCS; IRIS; OTTHUMP00000212147; OTTHUMP00000212148; OTTHUMP00000212149; OTTHUMP00000212150; OTTHUMP00000212151; OTTHUMP00000212155; PNCA4; PPP1R53; protein phosphatase 1, regulatory subunit 53; PSCP; RING finger protein 53; RING-type E3 ubiquitin transferase BRCA1; RNF53; truncated BRCA1; truncated breast and ovarian cancer susceptibility protein 1; truncated breast and ovarian cancer sususceptibility protein 1; truncated breast cancer type 1 susceptibility protein | |
Rabbit | |
sequential chromatography | |
RUO | |
12189, 497672, 672 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O54952, P38398, P48754 | |
BRCA1 | |
A synthesized peptide derived from human BRCA1(Accession P38398), corresponding to amino acid residues around phosphorylated Ser1524. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction