Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-c-Jun (Ser73) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540192

Catalog No. PA540192


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

Transcription factor that recognizes and binds to the enhancer heptamer motif 5 ft.-TGA[CG]TCA-3 ft.. Promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation.
Specifications

Specifications

Phospho-c-Jun (Ser73)
Polyclonal
Unconjugated
JUN
Liquid
JUN
Synthesized peptide derived from human AP-1 around the phosphorylation site of S73.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P05412
JUN; Transcription factor AP-1; Activator protein 1; AP1; Proto-oncogene c-Jun; V-jun avian sarcoma virus 17 oncogene homolog; p39; JUND; Transcription factor jun-D
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3725
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit