Learn More
Phospho-c-Myc (Ser62) Rabbit anti-Human, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Manufacturer:  Thermo Scientific MA533183
Catalog No. PIMA533183
Add to cart
Description
Description
The c-Myc protein is a transcription factor, which is encoded by the c-Myc gene on human chromosome 8q24. c-Myc is commonly activated in a variety of tumor cells and plays an important role in cellular proliferation, differentiation, apoptosis and cell cycle progression. The phosphorylation of c-Myc has been investigated and previous studies have suggested a functional association between phosphorylation at Thr58/Ser62 by glycogen synthase kinase 3, cyclin dependent kinase, ERK2 and C-Jun N terminal Kinase (JNK) in cell proliferation and cell cycle regulation. Studies also have shown that c-Myc is essential for tumor cell development in vasculogenesis and angiogenesis that distribute blood throughout the cells, and which brought extensive attention in the development of new therapeutic approach for cancer treatment.
Specifications
Specifications
Phospho-c-Myc (Ser62)
Recombinant Monoclonal
Unconjugated
MYC
Liquid
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4609
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
0.9 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P01106
MYC
A synthesized peptide derived from human Phospho-MYC (S62)
Protein G
RUO
Antibody
Monoclonal
Human