Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-c-Myc (Ser81) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$145.80 - $373.00

Specifications

Antigen phospho-c-Myc (Ser81)
Concentration 1.0 mg/mL
Applications Functional Assay, Inhibition Assays, Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABC1471100
View Documents
MilliporeSigma
ABC1471100UG
100 μg
Each for $373.00
Add to cart
 
ABC147125UG
View Documents
MilliporeSigma
ABC147125UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

phospho-c-Myc (Ser81) Polyclonal antibody specifically detects phospho-c-Myc (Ser81) in Human samples. It is validated for Affects Function, Peptide Inhibition Assay, Western Blotting.
Specifications

Specifications

phospho-c-Myc (Ser81)
Functional Assay, Inhibition Assays, Western Blot
Purified
RUO
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
1.0 mg/mL
Unconjugated
Rabbit
Human
KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserine 81 from human c-Myc.
Polyclonal
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit