Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Cdc25A (Ser75) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540196

Catalog No. PA540196


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Directly dephosphorylates CDK1 and stimulates its kinase activity. Also dephosphorylates CDK2 in complex with cyclin E, in vitro.
Specifications

Specifications

Phospho-Cdc25A (Ser75)
Polyclonal
Unconjugated
CDC25A
Liquid
CDC25A
Synthesized peptide derived from human Cdc25A around the phosphorylation site of S75.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P30304
CDC25A; M-phase inducer phosphatase 1; Dual specificity phosphatase Cdc25A
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
993
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit