Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Creatine Kinase MT (Tyr153) Polyclonal Antibody
Phospho-Creatine Kinase MT (Tyr153) Polyclonal Antibody
Supplier: Thermo Scientific PA595676
Description
Phospho-Creatine Kinase MT (Tyr153) Polyclonal Antibody for Western Blot

Specifications
Phospho-Creatine Kinase MT (Tyr153) | |
Polyclonal | |
Unconjugated | |
CKMT1A | |
acidic-type mitochondrial creatine kinase; CKMT; CKMT1; CKMT1A; CKMT1B; Creatine kinase U-type, mitochondrial; creatine kinase, mitochondrial 1 (ubiquitous); creatine kinase, mitochondrial 1, ubiquitous; creatine kinase, mitochondrial 1A; creatine kinase, mitochondrial 1B; mia-CK; mi-CK; Mt-CK; ScCKmit; UbCKmit; ubiquitous mitochondrial creatine kinase; UMTCK; U-MtCK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1159, 12716, 548596 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.25 mg/mL | |
10mM HEPES with 100μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5 | |
P12532, P30275 | |
Ckmt1, CKMT1A, CKMT1B | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr153 conjugated to KLH. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction