Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CrkL (Tyr207) Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540278

Catalog No. PA540278


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

May mediate the transduction of intracellular signals.
Specifications

Specifications

Phospho-CrkL (Tyr207)
Polyclonal
Unconjugated
CRKL
Liquid
CRKL
Synthesized peptide derived from human Crk-L around the phosphorylation site of Y207.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Non-human Primate
Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P46109
CRKL; Crk-like protein
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1399
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit