Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Crkl (Tyr251) Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  BiossSupplier Diversity Partner BS3738R

Catalog No. 50-198-5620


Add to cart

Description

Description

May mediate the transduction of intracellular signals.
Specifications

Specifications

Phospho-Crkl (Tyr251)
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CRKL
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
1399
CRKL
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
P46109
1110025F07Rik; AA589403; AI325100; CRK like proto-oncogene, adaptor protein; Crkl; crkl {ECO:0000312; Crk-like protein; crk-like protein {ECO:0000250; Crkol; EMBL:AAH85865.1, ECO:0000312; OTTHUMP00000198539; RGD:1308531}; UniProtKB:P46109}; v-crk avian sarcoma virus CT10 oncogene homolog-like; v-crk sarcoma virus CT10 oncogene homolog (avian)-like; v-crk sarcoma virus CT10 oncogene homolog-like
KLH conjugated synthetic phosphopeptide derived from human Crkl around the phosphorylation site of Tyr251 [CA (p-Y)D].
Protein A
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit