Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Cyclin D2 (Thr280) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA540166

Catalog No. PA540166


Add to Cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

Phospho-Cyclin D2 (Thr280) Polyclonal antibody specifically detects Phospho-Cyclin D2 (Thr280) in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-Cyclin D2 (Thr280)
Polyclonal
Unconjugated
CCND2
2600016F06Rik; AI256817; BF642806; C86853; CCND2; cD2; cyclin D2; Cyl-2; G1/S-specific cyclin-D2; KIAK0002; MGC102758; MPPH3; Vin1; Vin-1; Vin-1 proto-oncogene
Rabbit
Antigen affinity chromatography
RUO
12444, 64033, 894
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P30279, P30280, Q04827
CCND2
Peptide sequence around phosphorylation site of threonine 280 (A-S-T(p)-P-T) derived from Human Cyclin D2.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit