Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-delta 1 Catenin (Tyr96) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12994R

Description
Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.Specifications
Phospho-delta 1 Catenin (Tyr96) | |
Polyclonal | |
Unconjugated | |
CTNND1 | |
AA409437; AU019353; Cadherin-associated Src substrate; CAS; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; Catenin delta-1; catenin src; Catns; CTNND; CTNND1; Kiaa0384; mKIAA0384; P120; p120 catenin; P120 p120; P120 P120CTN; p120(cas); p120(ctn); P120CAS; p120-catenin; P120CTN; RCJMB04_21j12 | |
Rabbit | |
Protein A | |
RUO | |
1500 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O60716 | |
CTNND1 | |
KLH conjugated synthetic phosphopeptide derived from human delta 1 Catenin around the phosphorylation site of Tyr96. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction