Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-EphB1/EphB3/EphB4 (Tyr778, Tyr792, Tyr774) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105950
Description
Antibody detects endogenous levels of EPH B1/3/4 only when phosphorylated at Tyr778/792/774.
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
Specifications
Phospho-EphB1/EphB3/EphB4 (Tyr778, Tyr792, Tyr774) | |
Polyclonal | |
Unconjugated | |
EPHB1 | |
9330129L11; AI042935; AW456895; AW488255; C130099E04Rik; Cek10; Cek6; Developmental kinase 2; developmental kinase 5; EK2; EK6; Elk; Elkh; ELK-related protein tyrosine kinase; Embryonic kinase 2; ENSMUSG00000074119; EPH receptor B1; Eph receptor B2 (ELK-related protein tyrosine kinase); Eph receptor B3; Eph receptor B4; eph tyrosine kinase 2; Ephb1; Ephb2; EPHB3; Ephb4; EPH-like kinase 2; EPH-like kinase 6; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; ephrin receptor EphB4; ephrin type-B receptor 1; Ephrin type-B receptor 3; ephrin type-B receptor 4; EPHT2; Epth2; Erk; ETK2; HEK2; Hek6; Hepatoma transmembrane kinase; HTK; human embryo kinase 2; MDK2; mDK-2; Mdk5; mDK-5; Myk1; NET; Neuronally-expressed EPH-related tyrosine kinase; Sek4; soluble EPHB1 variant 1; soluble EPHB4 variant 1; soluble EPHB4 variant 2; soluble EPHB4 variant 3; Tyro11; TYRO6; Tyrosine kinase MYK-1; tyrosine-protein kinase receptor EPH-2; tyrosine-protein kinase receptor HTK; tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO11; tyrosine-protein kinase TYRO6 | |
Rabbit | |
Sequential chromatography | |
RUO | |
13845, 13846, 2047, 2049, 2050, 24338, 270190, 287989, 686310 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P09759, P54753, P54754, P54760, P54761, P54762, Q8CBF3 | |
EPHB1, EPHB3, EPHB4 | |
A synthesized peptide derived from human EPHB1(Accession P54762), corresponding to amino acid residues around phosphorylated Tyr778. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction