Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EpoR (Tyr426) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540305

Catalog No. PA540305


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

Receptor for erythropoietin. Mediates erythropoietin-induced erythroblast proliferation and differentiation. Upon EPO stimulation, EPOR dimerizes triggering the JAK2/STAT5 signaling cascade. In some cell types, can also activate STAT1 and STAT3. May also activate the LYN tyrosine kinase.
Specifications

Specifications

Phospho-EpoR (Tyr426)
Polyclonal
Unconjugated
EPOR
Liquid
EPOR
Synthesized peptide derived from human EpoR around the phosphorylation site of Y426.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P19235
EPOR; Erythropoietin receptor; EPO-R, pEpoR
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2057
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit