Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MAPK15 (Thr175, Tyr177) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540156

Catalog No. PA540156


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

In vitro, phosphorylates MBP.
Specifications

Specifications

Phospho-MAPK15 (Thr175, Tyr177)
Polyclonal
Unconjugated
MAPK15
Liquid
MAPK15
Peptide sequence around phosphorylation site of threonine 175 and tyrosine 177 (A-V-T(p)-E-Y(p)-V-A) derived from Human ERK8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8TD08
EC 2.7.11.24, Extracellular signal-regulated kinase 8
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
225689
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit