Learn More
Phospho-FAK (Tyr397) Monoclonal Antibody (M121), Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Scientific MA547626
Encompass
Catalog No. PIMA547626
Add to Cart
Description
Description
The antibody detects a 125 kDa* protein on SDS-PAGE immunoblots of untreated HUVEC cells. This phosphorylated band is greatly reduced after treatment with alkaline phosphatase.
Phospho-FAK (Tyr397) Monoclonal antibody specifically detects Phospho-FAK (Tyr397) in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-FAK (Tyr397)
Monoclonal
Lot-Specific
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Q14118, Q62165
Mouse
Protein A
RUO
114489, 13138, 1605
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
M121
Unconjugated
DAG1
CG10023; CG10023-PA; CG10023-PB; CG10023-PC; CG10023-PD; CG10023-PF; CG10023-PG; CT28129; DFAK; DFak56; Dmel\CG10023; Dmel_CG10023; DmFAK; EC 2.7.10.2; Fadk; FADK 1; Fak; Fak1; Fak56; fak56D; FAK65D; Fak-PA; Fak-PB; Fak-PC; Fak-PD; Fak-PF; Fak-PG; FAK-related non-kinase polypeptide; focal adhesion kinase; Focal adhesion kinase 1; focal adhesion kinase homolog; focal adhesion kinase isoform FAK-Del33; focal adhesion kinase pp125FAK; focal adhesion kinase-related nonkinase; focal ashension kinase 1; FRNK; I79_019430; Kiaa4203; mKIAA4203; p125FAK; p41/p43FRNK; pFAK; pp125 PTK2; pp125FAK; PPP1R71; Protein phosphatase 1 regulatory subunit 71; protein phosphatase 1, regulatory subunit 71; protein tyrosine kinase 2; protein tyrosine kinase 2 L homeolog; protein-tyrosine kinase; protein-tyrosine kinase 2; PTK2; PTK2 protein tyrosine kinase 2; ptk2.L; tyrosine kinase; XELAEV_18032408mg; XPFAK
Clone (M117) was generated from a synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 892 of human dystroglycan. This peptide sequence has high homology to the conserved tyrosine site in rat and mouse dystroglycan.
100 μL
Primary
Human, Mouse, Rat
Liquid
Recommendations
Recommendations