Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Filamin A (Ser1084) Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS13164R
Description
Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.Specifications
Phospho-Filamin A (Ser1084) | |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Flna | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
2316 |
Flna | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P21333 | |
450 kDa protein; ABP-280; ABPX; actin binding protein 280; Actin-binding protein 280; alpha filamin; Alpha-filamin; CSBS; CVD1; dilated pupils 2; Dilp2; Endothelial actin-binding protein; Endothelial actin-binding protein 280); F730004A14Rik; filamin 1; filamin A; filamin A, alpha; filamin, alpha; Filamin1; Filamin-1; FilaminA; Filamin-A; FLN; Fln1; Flna; FLN-A; FMD; GENA 379; MNS; NHBP; Non-muscle filamin; OPD; OPD1; OPD2; RGD1560614; XLVD; XMVD | |
KLH conjugated synthetic phosphopeptide derived from human FLNA around the phosphorylation site of Ser1084. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title