Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-FOXO1/FOXO3 (Ser322, Ser325) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA540217
Description
This antibody is predicted to react with human and rat based on sequence homology.
Phospho-FOXO1/FOXO3 (Ser322, Ser325) Polyclonal antibody specifically detects Phospho-FOXO1/FOXO3 (Ser322, Ser325) in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Phospho-FOXO1/FOXO3 (Ser322, Ser325) | |
Polyclonal | |
Unconjugated | |
FOXO1 | |
1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 protein; AFX; AFX1; Afxh; AI876417; C76856; FKH1; Fkhr; Fkhr1; Fkhr2; Fkhrl1; FKHRL1P2; forkhead box O1; forkhead box O1a; forkhead box O1A (rhabdomyosarcoma); forkhead box O3; forkhead box O3A; forkhead box protein O1; Forkhead box protein O1A; forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; Forkhead in rhabdomyosarcoma; Forkhead in rhabdomyosarcoma-like 1; forkhead protein 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO1; Foxo1a; FOXO2; FOXO3; Foxo3a; FOXO4; MGC120490; MLLT7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2308, 2309, 294515, 56458, 56484, 84482 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
G3V7R4, O43524, Q12778, Q9R1E0, Q9WVH4 | |
FOXO1, FOXO3 | |
Synthesized peptide derived from human FoxO1/3 around the phosphorylation site of S322/S325. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction