Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-GAB2 (Tyr643) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5118517

Description
Phospho-GAB2 (Tyr643) Polyclonal antibody specifically detects Phospho-GAB2 (Tyr643) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
Phospho-GAB2 (Tyr643) | |
Polyclonal | |
Unconjugated | |
GAB2 | |
AI463667; D130058I17Rik; GAB2; Grb2 associated binder 2; GRB2 associated binding protein 2; GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated-binding protein 2; GRB3-3; Growth factor receptor bound protein 2-associated protein 2; KIAA0571; p97; PH domain-containing adaptor molecule p97; pp100 | |
Peptide sequence around phosphorylation site of tyrosine 643(V-D-Y(p)-V-Q) derived from Human Gab2 . | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9EQH1, Q9UQC2, Q9Z1S8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14389, 84477, 9846 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title