Learn More
Phospho-Gsk3b (Ser389) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148501AP
Catalog No. 50-172-9776
Add to cart
Description
Description
Menin is a putative tumor suppressor associated with a syndrome known as multiple endocrine neoplasia type 1. In vitro studies have shown menin is localized to the nucleus, possesses two functional nuclear localization signals, and inhibits transcriptional activation by JunD, however, the function of this protein is not known. Two messages have been detected on northern blots but the larger message has not been characterized. Two variants of the shorter transcript have been identified where alternative splicing affects the CDS. Five variants where alternative splicing takes place in the 5 ft. UTR have also been identified.
Specifications
Specifications
Phospho-Gsk3b (Ser389)
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P18266, P49841, Q9WV60
7330414F15Rik, 8430431H08Rik, C86142, GSK 3, GSK 3beta, GSK3, Gsk3b
Rabbit
IgG
150 μL
-20°C
Primary
2932, 56637, 84027
Western Blot
Unconjugated
GSK3B
Liquid
GSK3B
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat