Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-HDAC1 (Ser406) Mouse anti-Human,Bovine,Monkey,Rat,Rabbit, KHL, Clone: BT-15, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen phospho-HDAC1 (Ser406)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Conjugate KHL
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE18211MI
View Documents
MilliporeSigma
MABE1821100UG
100 μg
Each for $386.00
Add to cart
 
MABE18212MI
View Documents
MilliporeSigma
MABE182125UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

phospho-HDAC1 (Ser406) Monoclonal antibody specifically detects phospho-HDAC1 (Ser406) in Human,Bovine,Monkey,Rat,Rabbit samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

phospho-HDAC1 (Ser406)
KHL
Mouse
Histone deacetylase 1;HD1
KLH-conjugated linear peptide corresponding to 19 amino acids surrounding phosphoserine 406 from the C-terminal region of human Histone deacetylase 1.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunocytochemistry, Immunoprecipitation, Western Blot
Purified
Human, Bovine, Monkey, Rat, Rabbit
HDAC1, RPD3L1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit