Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-Histone H1.5 (Thr10) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121453
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family.
Specifications
Phospho-Histone H1.5 (Thr10) | |
Polyclonal | |
Unconjugated | |
H1-5 | |
H1; H1 histone family; H1 histone family, member 5; H1.5; H1B; H1F5; H1s-3; HIST1H1B; histone 1, H1b; histone cluster 1, H1b; histone H1.5; Histone H1a; histone H1b; Histone H1s-3; MGC126630; MGC126632 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3009 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P16401 | |
H1-5, HIST1H1B | |
Synthesized peptide derived from Human Histone H1.5 protein (6-17aa). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction