Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H3 (Thr45) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540143

Catalog No. PA540143


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability.
Specifications

Specifications

Phospho-Histone H3 (Thr45)
Polyclonal
Unconjugated
HIST1H3A
Liquid
HIST1H3A
Peptide sequence around phosphorylation site of threonine 45(P-G-T(p)-V-A) derived from Human Histone H3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P68431
H3 histone antibody, HIST1H3A antibody, Histone cluster 1, H3a antibody
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8350
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit