Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho Histone H3 S10 Rabbit anti-Human,Mouse, KHL, Clone: 63-1C-8, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen phospho Histone H3 S10
Applications Dot Blot, Flow Cytometry, Immunocytochemistry, Inhibition Assays, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB122625MI
View Documents
MilliporeSigma
ZRB122625UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12264XMI
View Documents
MilliporeSigma
ZRB12264X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

phospho Histone H3 S10 Monoclonal antibody specifically detects phospho Histone H3 S10 in Human,Mouse samples. It is validated for Dot Blot, Flow Cytometry, Immunocytochemistry, Peptide Inhibition Assay, Western Blotting.
Specifications

Specifications

phospho Histone H3 S10
KHL
Rabbit
Histone H3/t;H3t;H3/g
KLH-conjugated linear peptide corresponding to 14 amino acids surrounding phosphoserine 10 of human Histone H3.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Dot Blot, Flow Cytometry, Immunocytochemistry, Inhibition Assays, Western Blot
Purified
Human, Mouse
HIST3H3, H3FT
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit