Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-HP1 beta (Thr51) Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA564831

Catalog No. PIPA564831


Add to cart

Description

Description

Phospho-HP1 beta (Thr51) Polyclonal Antibody detects endogenous levels of HP1 beta only when phosphorylated at Thr51.

This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family . The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The protein may play an important role in the epigenetic control of chromatin structure and gene expression. Several related pseudogenes are located on chromosomes 1, 3, and X. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications

Specifications

Phospho-HP1 beta (Thr51)
Polyclonal
Unconjugated
CBX1
Liquid
CBX1
A synthesized peptide derived from human HP1 beta (Phospho-Thr51)
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P83916, P83917
CBX
Rabbit
IgG
100 μL
-20°C
Primary
10951, 12412, 360609
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit