Learn More
Enzo Life SciencesSupplier Diversity Partner IkappaB alpha (pSer32/Ser36) monoclonal antibody (39A1413) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIKAMTF140E
Clone: 39A1413. Host: Mouse. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to a portion of human IκBα phosphorylated at Ser32/36. UniProt: P25963. Species Reactivity: Human, Mouse, Rat, Porcine, Dog, Bovine. Applications: IP, WB. Recommended dilutions: Immunoprecipitation (1-2µg/ml). Western Blot (1-2µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.05% BSA and 0.05% sodium azide.
Catalog No. 50-666-487