Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-IkB beta (Thr19) Polyclonal Antibody
Phospho-IkB beta (Thr19) Polyclonal Antibody
Supplier: Thermo Scientific PA536772
Description
Phospho-IkB beta (Thr19) Polyclonal Antibody for Western Blot

Specifications
Phospho-IkB beta (Thr19) | |
Polyclonal | |
Unconjugated | |
NFKBIB | |
I kappa B beta; I(Kappa)B(beta); ikappaBbeta; I-kappa-B-beta; IkB; IKB B; IKB beta; Ikbb; ikB-B; IkBbeta; ikB-beta; NF kappa BIB; NF-kappa-B inhibitor beta; NF-kappa-BIB; NFKB inhibitor beta; NFKBIB; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta; Thyroid receptor; thyroid receptor-interacting protein 9; TR-interacting protein 9; TRIP 9; TRIP9; TRIP-9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18036, 4793, 81525 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15653, Q60778, Q9JIA3 | |
NFKBIB | |
Synthetic phosphopeptide derived from human IkB-beta around the phosphorylation site of Threonine 19. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction