Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-IRS1 (Ser612) Polyclonal Antibody
Supplier: Thermo Scientific PA5117201
Description

Specifications
Phospho-IRS1 (Ser612) | |
Polyclonal | |
Unconjugated | |
IRS1 | |
G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16367, 25467, 3667 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35568, P35569, P35570 | |
IRS1 | |
A synthetic phosphorylated peptide around S612 of human IRS1 (NP_0055351). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction