Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-IRS1 (Ser639) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5118551

Description
Phospho-IRS1 (Ser639) Polyclonal antibody specifically detects Phospho-IRS1 (Ser639) in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
Phospho-IRS1 (Ser639) | |
Polyclonal | |
Unconjugated | |
IRS1 | |
G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
A synthesized peptide derived from human IRS1 around the phosphorylation site of Ser639. | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35568, P35569, P35570 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16367, 25467, 3667 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title