Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-JAK2 (Tyr1007, Tyr1008) Polyclonal Antibody
Supplier: Thermo Scientific PA5117229
Description

Specifications
Phospho-JAK2 (Tyr1007, Tyr1008) | |
Polyclonal | |
Unconjugated | |
JAK2 | |
EC 2.7.10.2; Fd17; JAK 2; JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); JTK 10; JTK10; kinase Jak2; THCYT3; Tyrosine-protein kinase JAK2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16452, 3717 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.48 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O60674, Q62120 | |
JAK2 | |
A synthetic phosphorylated peptide around Y1007 of human JAK2 (NP_0049631). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction