Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho JNK (Thr183 Rabbit anti-Human,Mouse, KHL, Clone: 3F7, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen phospho JNK (Thr183
Applications Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB117325MI
View Documents
MilliporeSigma
ZRB117325UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11734XMI
View Documents
MilliporeSigma
ZRB11734X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

phospho JNK (Thr183 Monoclonal antibody specifically detects phospho JNK (Thr183 in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Peptide Inhibition Assay, Western Blotting.
Specifications

Specifications

phospho JNK (Thr183
KHL
Rabbit
Mitogen-activated protein kinase 8/9;MAP kinase 8/9;MAPK8/9;Stress-activated protein kinase 1c/1a;SAPK1c/SAPK1a;JNK1/ JNK2;c-Jun N-terminal kinase 1/2
KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphothreonine 183 and phosphotyrosine 185 from human JNK/SAPK.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot
Purified
Human, Mouse
MAPK8/9, JNK1/JNK2, PRKM8/9, SAPK1/2, SAPK1C/1A
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit