Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-JunB (Thr102, Thr104) Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11228R

Description
JunB ,a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5′-TGA[CG]TCA-3′. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.Specifications
Phospho-JunB (Thr102, Thr104) | |
Polyclonal | |
Unconjugated | |
Junb | |
activator protein 1; AP-1; Jun B; jun B proto-oncogene; JUNB; Jun-b; Jun-B oncogene; JunB proto-oncogene, AP-1 transcription factor subunit; MyD21; Oncogene Jun-B; Transcription factor jun-B | |
Rabbit | |
Protein A | |
RUO | |
16477, 24517, 3726 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P09450, P17275, P24898 | |
Junb | |
KLH conjugated synthetic phosphopeptide derived from human JunB around the phosphorylation site of Thr102/Thr104 [TT (p-T)P (p-T)PP]. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction