Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Ku80 (Thr714) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538118
Description
XRCC5 encoded by this gene is the 80-kilodalton subunit of the Ku heterodimer protein which is also known as ATP-dependant DNA helicase II or DNA repair protein XRCC5. Ku is the DNA-binding component of the DNA-dependent protein kinase, and it functions together with the DNA ligase IV-XRCC4 complex in the repair of DNA double-strand break by non-homologous end joining and the completion of V(D)J recombination events. This gene functionally complements Chinese hamster xrs-6, a mutant defective in DNA double-strand break repair and in ability to undergo V(D)J recombination. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity.
Specifications
Phospho-Ku80 (Thr714) | |
Polyclonal | |
Unconjugated | |
XRCC5 | |
86 kDa subunit of Ku antigen, ATP-dependent DNA helicase II, 80 kDa subunit, CTC box binding factor 85 kDa subunit, CTC85, CTCBF, DNA-repair protein XRCC5, G22P2, KU86, Lupus Ku autoantigen protein p86, Nuclear factor IV, TLAA, Thyroid- lupus autoantigen, XRCC5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7520 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P13010 | |
XRCC5 | |
A synthetic phosphopeptide derived from human Ku80 around the phosphorylation site of Thr714 (G-D-TP-A-A) | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction