Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MAP3K12 (Ser298, Ser302) Rabbit anti-Human, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc PMAP3K12140AP
Description
Phospho-MAP3K12 (Ser298, Ser302) Polyclonal antibody specifically detects Phospho-MAP3K12 (Ser298, Ser302) in Human, Rat samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
Phospho-MAP3K12 (Ser298, Ser302) | |
Polyclonal | |
Unconjugated | |
MAP3K12 | |
DLK; Dual leucine zipper bearing kinase; dual leucine zipper kinase; dual leucine zipper kinase DLK; leucine zipper protein kinase; leucine-zipper protein kinase; M3KC; MAP3K12; MAPK-upstream kinase; MEKK12; Mitogen activated protein kinase 12 (Zipper (leucine) protein kinase); mitogen activated protein kinase kinase kinase 12; mitogen-activated protein kinase kinase kinase 12; Mixed lineage kinase; MUK; PK; protein kinase MUK; zipper (leucine) protein kinase; Zpk; ZPKP1 | |
Synthetic peptide taken within amino acid region 270-320 on MAP3K12 protein. Phosphorylated S298 & S302 residues. | |
200 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q12852, Q63796 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25579, 7786 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction