Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MLKL (Ser125) Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5105677

Catalog No. PIPA5105677


Only null left
Add to Cart

Description

Description

Antibody detects endogenous levels of MLKL only when phosphorylated at Ser125.

The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-MLKL (Ser125)
Polyclonal
Unconjugated
Mlkl
9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like
Rabbit
Sequential Chromatography
RUO
197259, 690743, 74568
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NB16, Q9D2Y4
Mlkl
A synthesized peptide derived from human MLKL(Accession Q8NB16), corresponding to amino acid residues around phosphorylated Ser125.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.