Learn More
Human Phospho-MSK1 (S376)/MSK2 (S360) Antibody, R&D Systems™
Rabbit Polyclonal Antibody
Supplier:  R&D Systems AF1094SP
Catalog No. AF1094SP
Add to Cart
This item is not returnable. View return policy
Description
Description
MSK1/MSK2 Polyclonal specifically detects MSK1/MSK2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
MSK1/MSK2 (p Ser376, p Ser360)
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Rabbit
Affinity Purified
RUO
Detects human MSK1 and MSK2 when phosphorylated at S376 and S360, respectively. The amino acid sequence surrounding S360 of mouse MSK2 is identical to the phosphopeptide antigen.
Human
IgG
Western Blot
LYOPH
Western Blot 0.2 ug/mL
1110069D02Rik, 3110005L17Rik, 6330404E13Rik, RPS6KA4, RPS6KA5, RSKB
Phosphopeptide containing human MSK1 S376 site
25 μg
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions