Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-mTOR (Ser2448) Monoclonal Antibody (2A12G3), CoraLite488, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL48867778100UL


This item is not returnable.
View return policy
Description
Phospho-mTOR (Ser2448) Monoclonal antibody specifically detects Phospho-mTOR (Ser2448) in Human, Mouse, Porcine, Rabbit, Rat samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
Phospho-mTOR (Ser2448) | |
Monoclonal | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O70325, P36968, P36969, P36970 | |
Mouse | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
IgG2b |
Flow Cytometry, Immunocytochemistry | |
2A12G3 | |
CoraLite 488 | |
GPX4 | |
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS | |
Protein A | |
RUO | |
2879, 29328, 399537, 625249 | |
Liquid | |
Human|Mouse|Porcine|Rabbit|Rat |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction