Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-NBS1 (Ser343) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578070
Description
Positive Control: HeLa, HeLa (100 J/M2 UVC treatment and recovery for 4 hr) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
Specifications
Phospho-NBS1 (Ser343) | |
Polyclonal | |
Unconjugated | |
NBN | |
AT V1; AT V2; ATV; AT-V1; AT-V2; Cell cycle regulatory protein p95; FLJ10155; I79_012607; MGC87362; Nbn; NBS; NBS1; nibrin; Nijmegen breakage syndrome 1 (nibrin); Nijmegen breakage syndrome protein 1; Nijmegen breakage syndrome protein 1 homolog; OTTHUMP00000204088; P95; p95 protein of the MRE11/RAD50 complex; p95-NBS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4683 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.97 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
O60934 | |
NBN | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human NBS1 (phospho Ser343). The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction