Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-NBS1 (Ser343) Rabbit anti-Human, Clone: SY0215, Invitrogen™

Rabbit Monoclonal Antibody

Manufacturer:  Thermo Scientific MA532093

Catalog No. PIMA532093


Add to cart

Description

Description

Rabbit Monoclonal Antibody

Phospho-NBS1 (Ser343) Monoclonal antibody specifically detects Phospho-NBS1 (Ser343) in Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Specifications

Specifications

Phospho-NBS1 (Ser343)
Monoclonal
1 mg/mL
TBS with 1% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
O60934
AT V1; AT V2; ATV; AT-V1; AT-V2; Cell cycle regulatory protein p95; FLJ10155; I79_012607; MGC87362; Nbn; NBS; NBS1; Nibrin; Nijmegen breakage syndrome protein 1; Nijmegen breakage syndrome protein 1 homolog; OTTHUMP00000204088; P95; p95-NBS1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4683
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
SY0215
Unconjugated
NBN
Liquid
NBN
Synthetic phospho-peptide corresponding to residues surrounding Ser343 of human p95/NBS1
Protein A
RUO
Antibody
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit