Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-NMDAR1 (Ser896) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA540256
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Phospho-NMDAR1 (Ser896) Polyclonal antibody specifically detects Phospho-NMDAR1 (Ser896) in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Phospho-NMDAR1 (Ser896) | |
Polyclonal | |
Unconjugated | |
GRIN1 | |
GluN1; Glun1r; GluRdelta1; Glurz1; GluRzeta1; glutamate [NMDA] receptor subunit zeta 1; glutamate [NMDA] receptor subunit zeta-1; glutamate ionotropic receptor NMDA type subunit 1; glutamate receptor; glutamate receptor ionotropic, NMDA 1; glutamate receptor subunit zeta 1; glutamate receptor, ionotropic, NMDA1 (zeta 1); glutamate receptor, ionotropic, N-methyl D-aspartate 1; GRIN1; M100174; MRD8; neurotransmitter receptor; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDA1; Nmdar; NMDAR1; NMD-R1; NMDZ1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor 1; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; NMZ1; NR1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; Rgsc174; RP11-350O14.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14810, 24408, 2902 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35438, P35439, Q05586 | |
GRIN1 | |
Synthesized peptide derived from human NMDAzeta1 around the phosphorylation site of S896. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction