Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-p53 (Ser376) Recombinant Rabbit Monoclonal Antibody (ST0440), Invitrogen™
Rabbit Monoclonal Antibody
Supplier: Thermo Scientific MA532142
Description
Rabbit Monoclonal Antibody
Phospho-p53 (Ser376) Monoclonal antibody specifically detects Phospho-p53 (Ser376) in Human samples. It is validated for Western BlotSpecifications
Phospho-p53 (Ser376) | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
P04637 | |
Tp53 | |
Synthetic phospho-peptide corresponding to residues surrounding Ser376 of human p53 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
ST0440 | |
Unconjugated | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; BOS_18908; Cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; OTTMUSP00000006194; p44; P53; Phosphoprotein p53; tp53; Trp248; Trp53; Tumor suppressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Rabbit | |
Protein A | |
RUO | |
7157 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction