Learn More
Phospho-p70 S6 Kinase Beta 2 (Tyr389) Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS3498R
Catalog No. 50-198-5449
Add to cart
Description
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates the S6 ribosomal protein and eucaryotic translation initiation factor 4B. Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
Specifications
Specifications
Phospho-p70 S6 Kinase Beta 2 (Tyr389)
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rps6kb2
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
6199
Rps6kb2
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q9UBS0
70 kDa ribosomal protein S6 kinase 2; 70kDa; KLS; p70 ribosomal S6 kinase beta; p70 S6 kinase beta; p70 S6KB; p70 S6K-beta; p70(S6K)-beta; P70-beta; P70-beta-1; P70-beta-2; p70-S6K 2; P70S6K2; p70S6Kb; ribosomal protein S6 kinase B2; Ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase, 70kD, polypeptide 2; ribosomal protein S6 kinase, 70kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; Rps6kb2; S6 kinase 2; S6 kinase-related kinase; S6K2; S6K-beta; S6K-beta2; S6K-beta-2; serine/threonine-protein kinase 14 beta; Serine/threonine-protein kinase 14B; SRK; STK14B
KLH conjugated synthetic phosphopeptide derived from human p70 S6 Kinase Beta 2 around the phosphorylation site of Tyr389.
Protein A
RUO
Antibody
Polyclonal
Human