Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-p73 (Ser388) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5118779

Description
Phospho-p73 (Ser388) Polyclonal antibody specifically detects Phospho-p73 (Ser388) in Human, Mouse samples. It is validated for Western BlotSpecifications
Phospho-p73 (Ser388) | |
Polyclonal | |
Unconjugated | |
TP73 | |
deltaNp73; OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; P73; RP23-254N4.10; TAp73; Tp73; transformation related protein 73; Trp73; Tumor protein p73 | |
A synthesized peptide derived from human p73 around the phosphorylation site of Ser388. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15350, Q9JJP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22062, 7161 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title