Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-PANX1 (Tyr198) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN1681
Description
Anti-Phospho-PANX1 (Tyr198) is a highly specific rabbit polyclonal antibody that targets Pannexin 1 and has been tested in Immunofluorescence and Western blotting.
Specifications
Phospho-PANX1 (Tyr198) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This polyclonal antibody specifically detects pannexin-1 phosphorylated on tyrosine 198 in HEK293 cells transfected with murine Pannexin 1. | |
Stable for 1 year at 2-8°C from date of receipt. |
Western Blot, Immunofluorescence | |
Please refer to lot specific datasheet. | |
Panx1 | |
A linear peptide corresponding to 12 amino acids surrounding phospho-Tyr198 of murine Pannexin-1. | |
100 μL | |
Neuroscience | |
NP_062355 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction